Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF280C Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA562419
Description
Immunogen sequence: VSKSSQSSVT VENASKPDFT KNSQVGSDNS SILLFDSTQE SLPPSQDIPA IFREGMKNTS YVLKHPSTSK VNSVTPKKPK TSEDV Highest antigen sequence identity to the following orthologs: Mouse - 50%, Rat - 47%.
May function as a transcription factor.
Specifications
ZNF280C | |
Polyclonal | |
Unconjugated | |
ZNF280C | |
SUHW3; Suppressor of hairy wing homolog 3; zinc finger protein 280C; Zinc finger protein 633; ZNF280C; ZNF633 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
55609 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8ND82 | |
ZNF280C | |
Recombinant protein corresponding to Human ZNF280C. Recombinant protein control fragment (Product #RP-101102). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction