Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF280C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101916
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101916 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101916 Supplier Invitrogen™ Supplier No. PA5101916
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total ZNF280C.

May function as a transcription factor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF280C
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ZNF280C
Gene Accession No. Q8ND82
Gene Alias SUHW3; Suppressor of hairy wing homolog 3; zinc finger protein 280C; Zinc finger protein 633; ZNF280C; ZNF633
Gene Symbols ZNF280C
Host Species Rabbit
Immunogen A synthesized peptide derived from human ZNF280C(Accession Q8ND82), corresponding to amino acid residues K253-E303.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55609
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.