Learn More
ZNF280D Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A303232AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
ZNF280D may function as a transcription factor.Specifications
ZNF280D | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q6N043 | |
ZNF280D | |
Between 879 and 929 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
ZNF280D | |
FLJ20086, SUHW4, suppressor of hairy wing homolog 4, suppressor of hairy wing homolog 4 (Drosophila), zinc finger protein 634, ZNF634 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
54816 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.