Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF281 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA521121
Description
A suggested positive control is A-20 cell lysate. PA5-21121 can be used with blocking peptide PEP-1235.
ZFP281, also known as ZBP-99, is a member of a conserved family of transcription factors that interact with GC-rich promoters. It contains four Kruppel-like zinc fingers and is highly homologous to ZBP-89, another regulator of ornithine decarboxylase gene expression. ZFP281 also interacts with many other other transcription factors, such as Sp1 to repress vimentin gene transcription. ZFP281 has also been shown to control embryonic stem cell pluripotency by direct regulating specific target genes. ZFP281 phyically interacts with the transcription factos POU5F1, Sox2, and NANOG. Gene expression microarray anlysis indicated that some ZFP281 target genes were activated, while others were repressed upon knockdown of ZFP281 expression, showing that ZFP281 plays bifuctinal roles in regulating gene expression in stem cells.
Specifications
ZNF281 | |
Polyclonal | |
Unconjugated | |
ZNF281 | |
GC-box-binding zinc finger protein 1; GZP1; Transcription factor ZBP-99; ZBP99; ZBP-99; Zfp281; Zinc finger DNA-binding protein 99; zinc finger protein 281; ZNF281; ZNP-99; ZNP-99 transcription factor | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
226442, 23528 | |
Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q99LI5, Q9Y2X9 | |
Zfp281, ZNF281 | |
A 19 amino acid peptide near the carboxy terminus of human ZFP281. | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction