Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF281 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562351
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562351 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562351 Supplier Invitrogen™ Supplier No. PA562351
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SSPLHNHTLF PEKQIYTTSP LECGFGQSVT SVLPSSLPKP PFGMLFGSQP GLYLSALDAT HQQLTPSQEL DDLIDSQKNL ETSSAFQSSS QKLTSQKEQ Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

ZFP281, also known as ZBP-99, is a member of a conserved family of transcription factors that interact with GC-rich promoters. It contains four Kruppel-like zinc fingers and is highly homologous to ZBP-89, another regulator of ornithine decarboxylase gene expression. ZFP281 also interacts with many other other transcription factors, such as Sp1 to repress vimentin gene transcription. ZFP281 has also been shown to control embryonic stem cell pluripotency by direct regulating specific target genes. ZFP281 phyically interacts with the transcription factos POU5F1, Sox2, and NANOG. Gene expression microarray anlysis indicated that some ZFP281 target genes were activated, while others were repressed upon knockdown of ZFP281 expression, showing that ZFP281 plays bifuctinal roles in regulating gene expression in stem cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF281
Applications ChIP Assay, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF281
Gene Accession No. Q9Y2X9
Gene Alias GC-box-binding zinc finger protein 1; GZP1; Transcription factor ZBP-99; ZBP99; ZBP-99; Zfp281; Zinc finger DNA-binding protein 99; zinc finger protein 281; ZNF281; ZNP-99; ZNP-99 transcription factor
Gene Symbols ZNF281
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZNF281. Recombinant protein control fragment (Product #RP-101420).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23528
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.