Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF282 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541464
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541464 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541464 Supplier Invitrogen™ Supplier No. PA541464
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: FIRKQNLLKH QRIHTGERPY TCGECGKSFR YKESLKDHLR VHSGGPGPGA Sequence homology: Cow: 100%; Dog: 79%; Guinea Pig: 75%; Horse: 79%; Human: 100%; Mouse: 100%; Pig: 93%; Rabbit: 100%; Rat: 100%; Zebrafish: 83%.

Binds to the U5 repressive element (U5RE) of the human T cell leukemia virus type I long terminal repeat. It recognizes the 5'-TCCACCCC-3' sequence as a core motif and exerts a strong repressive effect on HTLV-I LTR-mediated expression.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF282
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ZNF282
Gene Accession No. Q9UDV7
Gene Alias HTLV-I U5 repressive element-binding protein 1; HTLV-I U5RE-binding protein 1; HUB1; HUB-1; LOW QUALITY PROTEIN: zinc finger protein 282; zinc finger protein 282; ZNF282
Gene Symbols ZNF282
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human ZNF282 (aa 611-660).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8427
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.