Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF282 Polyclonal Antibody

Catalog No. PIPA555066
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555066 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555066 Supplier Invitrogen™ Supplier No. PA555066

Rabbit Polyclonal Antibody

Immunogen sequence: DIAVYFSEDE WKNLDEWQKE LYNNLVKENY KTLMSLDAEG SVPKPDAPVQ AEPREEPCVW EQRHPEEREI PMDPEAGAEP LVPAQDASSQ VKREDTLCVR GQRGLEERAI PTESITDSPI SAQDLLSRIK QEEHQCVWDQ QDLA Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 87%.

Binds to the U5 repressive element (U5RE) of the human T cell leukemia virus type I long terminal repeat. It recognizes the 5'-TCCACCCC-3' sequence as a core motif and exerts a strong repressive effect on HTLV-I LTR-mediated expression.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF282
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF282
Gene Accession No. Q9UDV7
Gene Alias HTLV-I U5 repressive element-binding protein 1; HTLV-I U5RE-binding protein 1; HUB1; HUB-1; LOW QUALITY PROTEIN: zinc finger protein 282; zinc finger protein 282; ZNF282
Gene Symbols ZNF282
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZNF282. Recombinant protein control fragment (Product #RP-89398).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8427
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.