Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF283 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567265
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567265 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567265 Supplier Invitrogen™ Supplier No. PA567265
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SQWEMKDKSKT LGLEASIFRN NWKCKSIFEG LKGHQEGYFS QMIISYEKIP SYRKSKSLTP HQ Highest antigen sequence identity to the following orthologs - mouse 37%, rat 33%.

May be involved in transcriptional regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF283
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF283
Gene Accession No. Q8N7M2
Gene Alias HZF19; HZF41; Zinc finger protein 283; zinc finger protein 41; zinc finger protein HZF19; ZNF283
Gene Symbols ZNF283
Host Species Rabbit
Immunogen Recombinant Human ZNF283. Recombinant protein control fragment (Product #RP-105575).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 284349
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.