Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF284 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541545
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541545 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541545 Supplier Invitrogen™ Supplier No. PA541545
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: KDVAVVFTEE ELGLLDVSQR KLYRDVMLEN FRNLLSVGHQ LSHRDTFHFQ Sequence homology: Cow: 93%; Dog: 86%; Horse: 85%; Human: 100%; Mouse: 85%; Pig: 93%; Rabbit: 92%; Rat: 86%.

ZNF284 may be involved in transcriptional regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF284
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ZNF284
Gene Accession No. Q2VY69
Gene Alias Zinc finger protein 284; ZNF284; ZNF284L
Gene Symbols ZNF284
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human ZNF284 (aa 11-60).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 342909
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.