Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF286 Antibody (OTI7D2), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274935H
Description
ZNF286 Monoclonal specifically detects ZNF286 in Human samples. It is validated for Western Blot.Specifications
ZNF286 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
ZNF286A | |
Recombinant protein fragment corresponding to amino acids 1-252 of human ZNF286 (NP_065703) produced in E.coli. | |
0.1 mL | |
57335.0 | |
Store at 4°C in the dark. |
Western Blot | |
OTI7D2 | |
Western Blot | |
MGC149627, zinc finger protein 286, zinc finger protein 286A | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction