Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF286A Monoclonal Antibody (OTI7D2)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA526602
Description
ZNF286A Monoclonal Antibody for Western Blot, IHC (P)
May be involved in transcriptional regulation.
Specifications
ZNF286A | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9HBT8 | |
ZNF286A | |
Human recombinant protein fragment corresponding to amino acids 1-252 of ZNF286A produced in E.coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
OTI7D2 | |
Unconjugated | |
ZNF286A | |
KIAA1874; Zinc finger protein 286A; zinc finger protein 624; zinc finger protein ZNF286; ZNF286; ZNF286A | |
Mouse | |
Affinity chromatography | |
RUO | |
57335 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction