Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF286A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA564989
Description
Immunogen sequence: ETDLAEMPEKG ALSSQDSPHF QEKSTEEGEV AALRLTARSQ ETVTFKDVAM DFTPEEWGKL DPAQR Highest antigen sequence identity to the following orthologs - mouse 76%, rat 80%.
May be involved in transcriptional regulation.
Specifications
ZNF286A | |
Polyclonal | |
Unconjugated | |
ZNF286A | |
KIAA1874; Zinc finger protein 286A; zinc finger protein 624; zinc finger protein ZNF286; ZNF286; ZNF286A | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
57335 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9HBT8 | |
ZNF286A | |
Recombinant Human ZNF286A. Recombinant protein control fragment (Product #RP-104570). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction