Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF287 Monoclonal Antibody (3E12)
GREENER_CHOICE

Catalog No. PIMA522352
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA522352 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA522352 Supplier Invitrogen™ Supplier No. MA522352
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: PEWETKAQAC TPVEDMSKLT KEETHTIKLE DSYDYDDRLE RRGKGGFWKI HTDERGFSLK SVLSQEYDPT EECLSKYDIY RNNFEKHSNL IVQFDTQLDN.

This gene encodes a member of the krueppel family of zinc finger proteins, suggesting a role as a transcription factor. Its specific function has not been determined. This gene is located near the Smith-Magenis syndrome region on chromosome 17.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF287
Applications ELISA, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 3E12
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene ZNF287
Gene Accession No. Q9HBT7
Gene Alias B230333C16Rik; mszf16; mszf74; Skat2; SKAT-2; Zfp287; Zfp-287; Zinc finger protein 287; zinc finger protein SKAT2; zinc finger protein SKAT-2; zinc finger protein with KRAB and SCAN domains 13; ZKSCAN13; Znf287; ZSCAN45
Gene Symbols ZNF287
Host Species Mouse
Immunogen ZNF287 (NP_065704.1, 231 a.a. ~ 330 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57336
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.