Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF287 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568882
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568882 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568882 Supplier Thermo Scientific Supplier No. PA568882
Add to Cart
Add to Cart

ZNF287 Polyclonal Antibody

ZNF287 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF287
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Zfp287
Gene Accession No. Q9EQB9
Gene Alias B230333C16Rik; mszf16; mszf74; Skat2; SKAT-2; Zfp287; Zfp-287; Zinc finger protein 287; zinc finger protein SKAT2; zinc finger protein SKAT-2; zinc finger protein with KRAB and SCAN domains 13; ZKSCAN13; Znf287; ZSCAN45
Gene Symbols Zfp287
Host Species Rabbit
Immunogen synthetic peptide directed towards the N terminal region of mouse ZNF287.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 170740
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.