Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF287 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA583078
Description
Immunogen sequence: PTVIPILEEP WMVIKEILEG PSPEWETKAQ ACTPVEDMSK LTKEETHTIK LEDSYDYDDR LERRGKGGFW KIHTDERGFS LKSVLSQEYD PTEECLSKYD IYRNNFEKHS NLIVQFDTQL DNKTSVYNEG RA.
This gene encodes a member of the krueppel family of zinc finger proteins, suggesting a role as a transcription factor. Its specific function has not been determined. This gene is located near the Smith-Magenis syndrome region on chromosome 17.
Specifications
ZNF287 | |
Polyclonal | |
Unconjugated | |
ZNF287 | |
B230333C16Rik; mszf16; mszf74; Skat2; SKAT-2; Zfp287; Zfp-287; Zinc finger protein 287; zinc finger protein SKAT2; zinc finger protein SKAT-2; zinc finger protein with KRAB and SCAN domains 13; ZKSCAN13; Znf287; ZSCAN45 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
57336 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9HBT7 | |
ZNF287 | |
Recombinant protein corresponding to Human ZNF287. Recombinant protein control fragment (Product #RP-99746). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction