Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF287 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA599298
Description
Antibody detects endogenous levels of total ZNF287.
This gene encodes a member of the krueppel family of zinc finger proteins, suggesting a role as a transcription factor. Its specific function has not been determined. This gene is located near the Smith-Magenis syndrome region on chromosome 17.
Specifications
ZNF287 | |
Polyclonal | |
Unconjugated | |
ZNF287 | |
B230333C16Rik; mszf16; mszf74; Skat2; SKAT-2; Zfp287; Zfp-287; Zinc finger protein 287; zinc finger protein SKAT2; zinc finger protein SKAT-2; zinc finger protein with KRAB and SCAN domains 13; ZKSCAN13; Znf287; ZSCAN45 | |
Rabbit | |
Affinity chromatography | |
RUO | |
170740, 57336 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q9EQB9, Q9HBT7 | |
Zfp287, ZNF287 | |
A synthesized peptide derived from human ZNF287(Accession Q9HBT7), corresponding to amino acid residues E244-S294. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction