Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF289/ARFGAP2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501984223
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-4223 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-4223 Supplier Bioss Supplier No. BS18504R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ARFGAP2 gene ontology annotations related to this gene include GTPase activator activity; metal ion binding.

Specifications

Antigen ZNF289/ARFGAP2
Applications Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Arfgap2
Gene Accession No. Q8N6H7
Gene Alias 2310032E02Rik; ADP ribosylation factor GTPase activating protein 2; ADP-ribosylation factor GTPase activating protein 2; ADP-ribosylation factor GTPase-activating protein 2; ARF GAP 2; ARFGAP2; GTPase-activating protein ZNF289; IRZ; Nbla10535; Zfp289; Zinc finger protein 289; zinc finger protein 289, ID1 regulated; Znf289
Gene Symbols Arfgap2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human ZNF289/ARFGAP2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84364
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.