Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF292 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA565628
Description
Immunogen sequence: NSLGTPSVPPK APVQKFSCQV EGCTRTYNSS QSIGKHMKTA HPDQYAAFKM QRKSKKGQKA NNLNTPNNGK FVYFLPSPVN SSNPFFTSQT KANGNPAC Highest antigen sequence identity to the following orthologs - mouse 86%, rat 83%.
May be involved in transcriptional regulation.
Specifications
ZNF292 | |
Polyclonal | |
Unconjugated | |
ZNF292 | |
16 zinc-finger domain protein; bA393I2.3; KIAA0530; Nbla00365; putative protein product of Nbla00365; ZFP292; Zinc finger protein 292; Zn-15; ZN-16; ZNF292 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23036 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.5 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O60281 | |
ZNF292 | |
Recombinant Human ZNF292. Recombinant protein control fragment (Product #RP-106296). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction