Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF294 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565958
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565958 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565958 Supplier Invitrogen™ Supplier No. PA565958
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ECLRFIMQQNL GEEEIEQMLV NDQLIPFIDA VLKDPGLQHG QLFNHLAETL SSWEAKADTE KDEKTAHNLE NVLI Highest antigen sequence identity to the following orthologs - mouse 75%, rat 77%.

ZNF294 may function as an E3 ubiquitin-protein ligase. E3 ubiquitin ligases accept ubiquitin from an E2 ubiquitin-conjugating enzyme in the form of a thioester and then directly transfer the ubiquitin to targeted substrates.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF294
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LTN1
Gene Accession No. O94822
Gene Alias 4930528H02Rik; AV266914; C21orf10; C21orf98; C87237; E3 ubiquitin-protein ligase listerin; HSPC087; KIAA0714; Lister; Listerin; listerin E3 ubiquitin protein ligase 1; LOC519526 protein; LOW QUALITY PROTEIN: E3 ubiquitin-protein ligase listerin; LTN1; RING finger protein 160; RING-type E3 ubiquitin transferase listerin; RNF160; Zfp294; zfp-294; Zinc finger protein 294; ZNF294
Gene Symbols LTN1
Host Species Rabbit
Immunogen Recombinant Human ZNF294. Recombinant protein control fragment (Product #RP-106647).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26046
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.