Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF294 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542315
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542315 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542315 Supplier Invitrogen™ Supplier No. PA542315
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: MGGKNKQRTK GNLRPSNSGR AAELLAKEQG TVPGFIGFGT SQSDLGYVPA Sequence homology: Cow: 100%; Dog: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 93%.

ZNF294 may function as an E3 ubiquitin-protein ligase. E3 ubiquitin ligases accept ubiquitin from an E2 ubiquitin-conjugating enzyme in the form of a thioester and then directly transfer the ubiquitin to targeted substrates.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF294
Applications Western Blot, Surface Plasmon Resonance
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene LTN1
Gene Accession No. O94822
Gene Alias 4930528H02Rik; AV266914; C21orf10; C21orf98; C87237; E3 ubiquitin-protein ligase listerin; HSPC087; KIAA0714; Lister; Listerin; listerin E3 ubiquitin protein ligase 1; LOC519526 protein; LOW QUALITY PROTEIN: E3 ubiquitin-protein ligase listerin; LTN1; RING finger protein 160; RING-type E3 ubiquitin transferase listerin; RNF160; Zfp294; zfp-294; Zinc finger protein 294; ZNF294
Gene Symbols LTN1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human ZNF294 (aa 1-50).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26046
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.