Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF295 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP220986
Description
ZNF295 Polyclonal specifically detects ZNF295 in Human samples. It is validated for Western Blot.Specifications
ZNF295 | |
Polyclonal | |
Unconjugated | |
0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
MGC15716, Zfp414, zinc finger protein 414 | |
Rabbit | |
119 kDa | |
0.1 mL | |
Primary | |
Human. Species reactivity based on sequence identity: Rat (82%). | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Western Blot | |
0.67mg/mL | |
Western Blot 1:500-1:3000 | |
Q9ULJ3 | |
ZNF295 | |
Recombinant protein encompassing a sequence within the N-terminus region of human ZNF295. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
49854 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction