Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF295 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA530607
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA530607 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA530607 Supplier Invitrogen™ Supplier No. PA530607
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A431. Predicted reactivity: Mouse (83%), Bovine (89%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

ZNF295 contains 1 BTB (POZ) domain and 8 C2H2-type zinc fingers. ZNF295 may be involved in transcriptional regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF295
Applications Western Blot
Classification Polyclonal
Concentration 0.67 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene ZBTB21
Gene Accession No. Q9ULJ3
Gene Alias 5430437K12Rik; B430213I24Rik; KIAA1227; mKIAA1227; ZBTB21; Zfp295; zinc finger and BTB domain containing 21; zinc finger and BTB domain-containing protein 21; Zinc finger protein 295; ZNF295
Gene Symbols ZBTB21
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the N-terminus region of human ZNF295. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 49854
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.