Learn More
Invitrogen™ ZNF295 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA555141
Description
Immunogen sequence: QPEPNKVNHI VTTKDDNVFS DSSEQVNFDS EDSSCLPEDL SLSKQLKIQV KEEPVEEAEE EAPEASTAPK EAGPSKEASL WPCEKCGKMF TVHKQLERHQ ELLCSVKPFI CHVCNKAFRT NFRLWSHFQS HMSQASEESA HKESEVCP Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 86%.
ZNF295 contains 1 BTB (POZ) domain and 8 C2H2-type zinc fingers. ZNF295 may be involved in transcriptional regulation.
Specifications
ZNF295 | |
Polyclonal | |
Unconjugated | |
ZBTB21 | |
5430437K12Rik; B430213I24Rik; KIAA1227; mKIAA1227; ZBTB21; Zfp295; zinc finger and BTB domain containing 21; zinc finger and BTB domain-containing protein 21; Zinc finger protein 295; ZNF295 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
49854 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9ULJ3 | |
ZBTB21 | |
Recombinant protein corresponding to Human ZNF295. Recombinant protein control fragment (Product #RP-100680). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.