Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF296 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA565567
Description
Immunogen sequence: MSRRKAGSAPR RVEPAPAANP DDEMEMQDLV IELKPEPDAQ PQQAPRLGPF SPKEVSS Highest antigen sequence identity to the following orthologs - mouse 36%, rat 47%.
May be a transcriptional corepressor with KLF4.
Specifications
ZNF296 | |
Polyclonal | |
Unconjugated | |
ZNF296 | |
ZFP296; zinc finger protein 296; Zinc finger protein 342; Znf296; ZNF342 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
162979 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8WUU4 | |
ZNF296 | |
Recombinant Human ZNF296. Recombinant protein control fragment (Product #RP-104874). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction