Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF297B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598248
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598248 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598248 Supplier Invitrogen™ Supplier No. PA598248
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ZNF297B Polyclonal Antibody for Western Blot, IHC (P), ELISA

ZNF297B is a candidate transcription factor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF297B
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene ZBTB43
Gene Accession No. O43298, Q9DAI4
Gene Alias 1700010E06Rik; Kiaa0414; mKIAA0414; ortholog of zinc finger protein 297B ZNF297B; ZBTB22B; ZBTB43; Zfp297b; zinc finger and BTB domain containing 43; zinc finger and BTB domain-containing protein 22B; Zinc finger and BTB domain-containing protein 43; zinc finger protein 297B; Znf297b; ZNF-X
Gene Symbols ZBTB43
Host Species Rabbit
Immunogen Recombinant Human Zinc finger and BTB domain-containing protein 43 protein (165-467AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23099, 71834
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.