Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF297B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA540428
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA540428 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA540428 Supplier Invitrogen™ Supplier No. PA540428
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: LVESFELGSG GHTDFPKAQE LRDGENEEES TKDELSSQLT EHEYLPSNSS Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 92%; Horse: 100%; Human: 100%; Mouse: 92%; Pig: 100%; Rabbit: 100%; Rat: 100%.

ZNF297B is a candidate transcription factor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF297B
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ZBTB43
Gene Accession No. O43298
Gene Alias 1700010E06Rik; Kiaa0414; mKIAA0414; ortholog of zinc finger protein 297B ZNF297B; ZBTB22B; ZBTB43; Zfp297b; zinc finger and BTB domain containing 43; zinc finger and BTB domain-containing protein 22B; Zinc finger and BTB domain-containing protein 43; zinc finger protein 297B; Znf297b; ZNF-X
Gene Symbols ZBTB43
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human ZNF297B (aa 151-200).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23099
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.