Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA540727
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA540727 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA540727 Supplier Invitrogen™ Supplier No. PA540727
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: METQADLVSQ EPQALLDSAL PSKVPAFSDK DSLGDEMLAA ALLKAKSQEL Sequence homology: Cow: 92%; Dog: 100%; Goat: 92%; Horse: 92%; Human: 100%; Mouse: 100%; Pig: 92%; Rat: 100%.

ZNF3 belongs to the krueppel C2H2-type zinc-finger protein family. It contains 8 C2H2-type zinc fingers and 1 KRAB domain. ZNF3 is involved in cell differentiation and/or proliferation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ZNF3
Gene Accession No. P17036
Gene Alias A8-51; AI854635; B230309J24; C2-H2 type zinc finger protein; Fnp-1; HF.12; KOX25; LOW QUALITY PROTEIN: zinc finger protein 3; MGC128512 protein; PP838; RGD1565881; Zfp113; Zfp3; Zfp-3; zinc finger and SCAN domain containing 21; zinc finger protein 3; zinc finger protein 3 homolog; zinc finger protein HF.12; Zinc finger protein HZF3.1; zinc finger protein KOX25; ZNF3; ZSCAN21
Gene Symbols ZNF3
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human ZNF3 (aa 1-50).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7551
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.