Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551989
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551989 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551989 Supplier Invitrogen™ Supplier No. PA551989
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SALPSKVPAF SDKDSLGDEM LAAALLKAKS QELVTFEDVA VYFIRKEWKR LEPAQRDLYR DVMLENYGNV FSLDRETRTE NDQEISEDTR SHGVLLGRFQ KDISQGLKFK EAYEREVSLK RPLGNSPGER LNRK Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 78%.

ZNF3 belongs to the krueppel C2H2-type zinc-finger protein family. It contains 8 C2H2-type zinc fingers and 1 KRAB domain. ZNF3 is involved in cell differentiation and/or proliferation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF3
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF3
Gene Accession No. P17036
Gene Alias A8-51; AI854635; B230309J24; C2-H2 type zinc finger protein; Fnp-1; HF.12; KOX25; LOW QUALITY PROTEIN: zinc finger protein 3; MGC128512 protein; PP838; RGD1565881; Zfp113; Zfp3; Zfp-3; zinc finger and SCAN domain containing 21; zinc finger protein 3; zinc finger protein 3 homolog; zinc finger protein HF.12; Zinc finger protein HZF3.1; zinc finger protein KOX25; ZNF3; ZSCAN21
Gene Symbols ZNF3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZNF3. Recombinant protein control fragment (Product #RP-99749).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7551
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.