Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF3 Polyclonal Antibody
Supplier: Thermo Scientific PA5113628
Description

Specifications
ZNF3 | |
Polyclonal | |
Unconjugated | |
ZNF3 | |
A8-51; AI854635; B230309J24; C2-H2 type zinc finger protein; Fnp-1; HF.12; KOX25; LOW QUALITY PROTEIN: zinc finger protein 3; MGC128512 protein; PP838; RGD1565881; Zfp113; Zfp3; Zfp-3; zinc finger and SCAN domain containing 21; zinc finger protein 3; zinc finger protein 3 homolog; zinc finger protein HF.12; Zinc finger protein HZF3.1; zinc finger protein KOX25; ZNF3; ZSCAN21 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
7551 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P17036 | |
ZNF3 | |
Synthetic peptide directed towards the N terminal region of human ZFP3. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction