Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF300 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
| Antigen | ZNF300 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
ZNF300 Polyclonal specifically detects ZNF300 in Human samples. It is validated for Western Blot.Specifications
| ZNF300 | |
| Polyclonal | |
| Rabbit | |
| NP_443092 | |
| 91975 | |
| Synthetic peptide directed towards the N terminal of human ZNF300. Peptide sequence DISNWIYPDEYQADGRQDRKSNLHNSQSCILGTVSFHHKILKGVTRDGSL. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| kruppel-like zinc finger protein, zinc finger protein 300 | |
| ZNF300 | |
| IgG | |
| 69 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title