Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF300 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA562480
Description
Immunogen sequence: EEPWIIKGDI SNWIYPDEYQ ADGRQDRKSN LHNSQSCILG TVSFHHKILK GVTRDGSLCS Highest antigen sequence identity to the following orthologs: Mouse - 28%, Rat - 32%.
The protein encoded by this gene is a C2H2-type zinc finger DNA binding protein and likely transcriptional regulator. The function of this protein is not yet known. Three transcript variants encoding different isoforms have been found for this gene.
Specifications
ZNF300 | |
Polyclonal | |
Unconjugated | |
ZNF300 | |
kruppel-like zinc finger protein; zinc finger protein 300; ZNF300 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
91975 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96RE9 | |
ZNF300 | |
Recombinant protein corresponding to Human ZNF300. Recombinant protein control fragment (Product #RP-101344). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction