Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF302 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5144777
Description
Immunogen sequence: KAYPFPLSHS VPASVNFGFS ALFEHCSEVT EIFELSELCV FWVLHFLSNS PNSTVEA Antigen sequence identity: Mouse 30%; Rat 28%.
May be involved in transcriptional regulation.
Specifications
ZNF302 | |
Polyclonal | |
Unconjugated | |
ZNF302 | |
HSD16; MST154; MSTP154; Zinc finger protein 135-like; Zinc finger protein 140-like; Zinc finger protein 302; zinc finger protein 327; ZNF135L; ZNF140L; ZNF302; ZNF327 | |
Rabbit | |
Affinity chromatography | |
RUO | |
55900 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid | |
IgG |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9NR11 | |
ZNF302 | |
Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 71-127. Recombinant protein control fragment (Product #RP-106942). | |
100 μL | |
Primary | |
Human | |
Antibody | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction