Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF302 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566260
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566260 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566260 Supplier Invitrogen™ Supplier No. PA566260
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KAYPFPLSHSV PASVNFGFSA LFEHCSEVTE IFELSELCVF WVLHFLSNSP NSTVEA Highest antigen sequence identity to the following orthologs - mouse 30%, rat 28%.

May be involved in transcriptional regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF302
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF302
Gene Accession No. Q9NR11
Gene Alias HSD16; MST154; MSTP154; Zinc finger protein 135-like; Zinc finger protein 140-like; Zinc finger protein 302; zinc finger protein 327; ZNF135L; ZNF140L; ZNF302; ZNF327
Gene Symbols ZNF302
Host Species Rabbit
Immunogen Recombinant protein fragment of Human ZNF302 (Product #RP-106942)
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55900
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.