Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF304 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA562207
Description
Immunogen sequence: SLRQHQGDYD GQMLFSCGDE GKAFLDTFTL LDSQMTHAEV RPFRCLPCGN VFKEKS Highest antigen sequence identity to the following orthologs: Mouse - 43%, Rat - 43%.
ZNF304 acts as transcriptional regulator and plays a role in gene silencing. It probably forms a corepressor complex required for activated KRAS-mediated promoter hypermethylation and transcriptional silencing of several tumor suppressor genes (TSGs) or other tumor-related genes in colorectal cancer (CRC) cells. ZNF304 is also required to maintain a transcriptionally repressive state of genes in undifferentiated embryonic stem cells (ESCs) by inducing trimethylation of Lysine-27 of histone H3 (H3K27me3) in a Polycomb group (PcG) complexes-dependent manner. The protein associates at promoter regions of TSGs and mediates the recruitment of the corepressor complex containing the scaffolding protein TRIM28, methyltransferase DNMT1 and histone methyltransferase SETDB1 and/or the PcG complexes at those sites. It acts as a transcription factor involved in the metastatic cascade process by inducing cell migration and proliferation and gain resistance to anoikis of ovarian carcinoma (OC) cells via integrin-mediated signaling pathways. It associates with the ITGB1 promoter and positively regulates beta-1 integrin transcription expression. It promotes angiogenesis and tumor growth.
Specifications
ZNF304 | |
Polyclonal | |
Unconjugated | |
ZNF304 | |
KRAB-containing zinc finger protein; zinc finger protein 304; ZNF304 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57343 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.5 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9HCX3 | |
ZNF304 | |
Recombinant protein corresponding to Human ZNF304. Recombinant protein control fragment (Product #RP-101284). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction