Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF304 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5107319
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5107319 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5107319 Supplier Invitrogen™ Supplier No. PA5107319
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Sequence homology: Cow: 77%; Dog: 100%; Horse: 92%; Human: 100%; Mouse: 77%; Pig: 77%; Rat: 93%.

ZNF304 acts as transcriptional regulator and plays a role in gene silencing. It probably forms a corepressor complex required for activated KRAS-mediated promoter hypermethylation and transcriptional silencing of several tumor suppressor genes (TSGs) or other tumor-related genes in colorectal cancer (CRC) cells. ZNF304 is also required to maintain a transcriptionally repressive state of genes in undifferentiated embryonic stem cells (ESCs) by inducing trimethylation of Lysine-27 of histone H3 (H3K27me3) in a Polycomb group (PcG) complexes-dependent manner. The protein associates at promoter regions of TSGs and mediates the recruitment of the corepressor complex containing the scaffolding protein TRIM28, methyltransferase DNMT1 and histone methyltransferase SETDB1 and/or the PcG complexes at those sites. It acts as a transcription factor involved in the metastatic cascade process by inducing cell migration and proliferation and gain resistance to anoikis of ovarian carcinoma (OC) cells via integrin-mediated signaling pathways. It associates with the ITGB1 promoter and positively regulates beta-1 integrin transcription expression. It promotes angiogenesis and tumor growth.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF304
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide; pH 7.4
Gene ZNF304
Gene Accession No. Q9HCX3
Gene Alias KRAB-containing zinc finger protein; zinc finger protein 304; ZNF304
Gene Symbols ZNF304
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N terminal region of human ZNF304 (aa 1-51).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57343
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.