Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF306 Monoclonal Antibody (2A1)
GREENER_CHOICE

Catalog No. PIMA522646
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA522646 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA522646 Supplier Invitrogen™ Supplier No. MA522646
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: CGKAFRRSSH LLRHQRIHTG DKNVQEPEQG EAWKSRMESQ LENVETPMSY KCNECERSFT QNTGLIEHQK IHTGEKPYQC NACGKGFTRI SYLVQHQRSH VGKNIL.

ZNF306 is a protein coding gene. Transcriptional factor that binds to the consensus sequence 5'-[GT][AG][AGT]GGGG-3' and acts as a repressor of autophagy. Specifically represses expression of genes involved in autophagy and lysosome biogenesis/function such as MAP1LC3B, ULK1 or WIPI2. Associates with chromatin at the ITGB4 and VEGF promoters. Also acts as a transcription activator and promotes cancer cell progression and/or migration in various tumors and myelomas.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF306
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2A1
Concentration 0.2-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene ZKSCAN3
Gene Accession No. Q9BRR0
Gene Alias 2810435N07Rik; AA408594; AI132359; dJ874C20.1; dJ874C20.1.; mszf35; SCAN-KRAB-zinc finger gene 1; SCAN-KRAB-zinc finger protein; Skz1; Zf47; ZFP306; Zfp307; Zfp47; zfp-47; Zinc finger and SCAN domain-containing protein 13; zinc finger protein 306; Zinc finger protein 307; zinc finger protein 309; Zinc finger protein 47 homolog; zinc finger protein with KRAB and SCAN domains 3; zinc finger protein zfp47; zinc finger with KRAB and SCAN domains 3; Zkscan3; ZNF306; ZNF309; ZSCAN13; ZSCAN35
Gene Symbols ZKSCAN3
Host Species Mouse
Immunogen ZKSCAN3 (NP_077819, 431 a.a. ~ 536 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80317
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.