Learn More
Invitrogen™ ZNF306 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA552671
Description
Immunogen sequence: LLTPAPGSQS SQFQLMKALL KHESVGSQPL QDRVLQVPVL AHGGCCREDK VVASRLTPES QGLLKVEDVA LTLTPEWTQQ DSSQGNLCRD EKQENHGSLV SLGDEKQTKS RDLPPAEELP EKEHGKISCH LREDIAQIPT CAEAGEQEGR Highest antigen sequence identity to the following orthologs: Mouse - 55%, Rat - 59%.
ZNF306 is a protein coding gene. Transcriptional factor that binds to the consensus sequence 5'-[GT][AG][AGT]GGGG-3' and acts as a repressor of autophagy. Specifically represses expression of genes involved in autophagy and lysosome biogenesis/function such as MAP1LC3B, ULK1 or WIPI2. Associates with chromatin at the ITGB4 and VEGF promoters. Also acts as a transcription activator and promotes cancer cell progression and/or migration in various tumors and myelomas.
Specifications
| ZNF306 | |
| Polyclonal | |
| Unconjugated | |
| ZKSCAN3 | |
| 2810435N07Rik; AA408594; AI132359; dJ874C20.1; dJ874C20.1.; mszf35; SCAN-KRAB-zinc finger gene 1; SCAN-KRAB-zinc finger protein; Skz1; Zf47; ZFP306; Zfp307; Zfp47; zfp-47; Zinc finger and SCAN domain-containing protein 13; zinc finger protein 306; Zinc finger protein 307; zinc finger protein 309; Zinc finger protein 47 homolog; zinc finger protein with KRAB and SCAN domains 3; zinc finger protein zfp47; zinc finger with KRAB and SCAN domains 3; Zkscan3; ZNF306; ZNF309; ZSCAN13; ZSCAN35 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 80317 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9BRR0 | |
| ZKSCAN3 | |
| Recombinant protein corresponding to Human ZNF306. Recombinant protein control fragment (Product #RP-89451). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.