Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF306 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5102903
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5102903 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5102903 Supplier Invitrogen™ Supplier No. PA5102903
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total ZKSCAN3.

ZNF306 is a protein coding gene. Transcriptional factor that binds to the consensus sequence 5'-[GT][AG][AGT]GGGG-3' and acts as a repressor of autophagy. Specifically represses expression of genes involved in autophagy and lysosome biogenesis/function such as MAP1LC3B, ULK1 or WIPI2. Associates with chromatin at the ITGB4 and VEGF promoters. Also acts as a transcription activator and promotes cancer cell progression and/or migration in various tumors and myelomas.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF306
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ZKSCAN3
Gene Accession No. Q91VW9, Q9BRR0
Gene Alias 2810435N07Rik; AA408594; AI132359; dJ874C20.1; dJ874C20.1.; mszf35; SCAN-KRAB-zinc finger gene 1; SCAN-KRAB-zinc finger protein; Skz1; Zf47; ZFP306; Zfp307; Zfp47; zfp-47; Zinc finger and SCAN domain-containing protein 13; zinc finger protein 306; Zinc finger protein 307; zinc finger protein 309; Zinc finger protein 47 homolog; zinc finger protein with KRAB and SCAN domains 3; zinc finger protein zfp47; zinc finger with KRAB and SCAN domains 3; Zkscan3; ZNF306; ZNF309; ZSCAN13; ZSCAN35
Gene Symbols ZKSCAN3
Host Species Rabbit
Immunogen A synthesized peptide derived from human ZKSCAN3(Accession Q9BRR0), corresponding to amino acid residues G212-D262.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 72739, 80317
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.