Learn More
Invitrogen™ ZNF31 Monoclonal Antibody (2C7)

Description
Peptide Sequence: MAMALELQAQ ASPQPEPEEL LIVKLEEDSW GSESKLWEKD RGSVSGPEAS RQRFRQFQYR DAAGPHEAFS QLWALCCRWL RPEIRLKEQI LELLVLEQFL TILPREVQTW VQARHPESGE EAVALVEDWH RETRTAGQSG LELHTEETRP LKTGEEAQSF QLQPVDPWPE GQSQKKGVKN TCPDLPNHLN AEVAPQPLKE SGVPVSKPSN TSEKEQGPEF WGLSLINSGK RSTADYSLDN EPAQALTWRD SRAWEEQYQW DVEDMKVSGV HWGYEETKTF LAILSESPFS EKLRTCHQNR QVYRAIAEQL RARGFLRTLE QCRYRVKNLL RNYRKAKSSH PPGTCPFYEE LEALVRARTA IRATDGPGEA VALPRLGDSD AEMDEQEEGG WDPEEMAEDC NGAGLVNVES TQGPRIAGAP ALFQSRIGKN MGV.
Specifications
Specifications
| Antigen | ZNF31 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 2C7 |
| Concentration | 0.2-1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | ZSCAN20 |
| Gene Accession No. | P17040 |
| Gene Alias | C130001F22; KOX29; Zfp31; ZFP-31; zinc finger and SCAN domain containing 20; zinc finger and SCAN domain-containing protein 20; zinc finger and SCAN domains 20; zinc finger protein 31; Zinc finger protein 360; zinc finger protein KOX29; zinc finger with KRAB and SCAN domains 20; Zkscan20; ZNF31; ZNF360; ZSCAN20 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.