Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF312 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541588
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541588 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541588 Supplier Invitrogen™ Supplier No. PA541588
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: THNDKKPFTC ATCGKGFCRN FDLKKHVRKL HDSVGPAAPS AKDLTRTVQS Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 93%.

ZNF312 is a candidate transcription factor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF312
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene FEZF2
Gene Accession No. Q8TBJ5
Gene Alias AI451466; AI852056; cadps; fez; Fez family zinc finger 2; fez family zinc finger protein 2; fezf2; Fezl; fezl protein; FKSG36; forebrain embryonic zinc finger; forebrain embryonic zinc finger-like protein; forebrain embryonic zinc finger-like protein 2; forebrain embryonic zinc-finger like; foreheadin; mutant embryonic zinc finger-like protein; SI:dZ6A2.1; SI:dZ6A2.7; testis tissue sperm-binding protein Li 80P; TOF; too few; Zfp312; zgc:92690; zinc finger protein; zinc finger protein 312; Zinc finger protein Fez-like; zinc-finger protein; ZNF312
Gene Symbols FEZF2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human ZNF312 (aa 410-459).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55079
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.