Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF313 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554344
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554344 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554344 Supplier Invitrogen™ Supplier No. PA554344
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SALAPGVRAV ELERQIESTE TSCHGCRKNF FLSKIRSHVA TCSKYQNYIM EGVKATIKDA SLQPRNVPNR YTFPCPYCPE KNFDQEGLVE HCKLFHSTDT KSVVCPICAS MPW Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 91%.

ZNF313 is a novel human zinc finger protein gene that contains both ring finger and C(2)H(2) domain. It may play a role in spermatogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF313
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RNF114
Gene Accession No. Q6J2U6, Q9ET26, Q9Y508
Gene Alias 1110008J21Rik; AI225886; AW549494; E3 ubiquitin-protein ligase RNF114; PSORS12; RING finger protein 114; RING-type E3 ubiquitin transferase RNF114; RNF114; Zfp228; Zfp313; Zinc finger protein 228; zinc finger protein 313; ZNF228; Znf313
Gene Symbols RNF114
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZNF313. Recombinant protein control fragment (Product #RP-91628).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 362277, 55905, 81018
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.