Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF318 Antibody, DyLight 650, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10077289C
Description
ZNF318 Polyclonal antibody specifically detects ZNF318 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
ZNF318 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
Zinc finger and SCAN domain-containing protein 15, zinc finger protein 397, Zinc finger protein 47ZSCAN15MGC13250, ZNF47 | |
The immunogen recognized by this antibody maps to a region between residue 2049 and 2099 of human zinc finger protein 318 (testicular zinc finger) using the numbering given in entry AAD17298.1 (GeneID 24149). | |
0.1 mL | |
Chromatin Research | |
24149 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
DyLight 650 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction