Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF318 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981346
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1346 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1346 Supplier Bioss Supplier No. BS11414R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ZNF318 encodes a nuclear protein with a zinc finger motif of the Cys2-His2 type that is a novel corepressor of androgen receptor (AR).

Specifications

Antigen ZNF318
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene ZNF318
Gene Accession No. Q5VUA4
Gene Alias 2610034E08Rik; D530032D06Rik; endocrine regulator; Endocrine regulatory protein; HRIHFB2436; testicular zinc finger; Testicular zinc finger protein; Tzf; Zfp318; Zinc finger protein 318; Znf318
Gene Symbols ZNF318
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human ZNF318.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24149
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.