Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF318 Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

Supplier:  Novus Biologicals NBP310946100UL

 View more versions of this product

Catalog No. NB126791


Only null left
Add to Cart

Description

Description

ZNF318 Polyclonal specifically detects ZNF318 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

ZNF318
Polyclonal
Western Blot 1.0 ug/ml, Immunohistochemistry
Zinc finger and SCAN domain-containing protein 15, zinc finger protein 397, Zinc finger protein 47ZSCAN15MGC13250, ZNF47
The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF318 (NP_055160). Peptide sequence SVFTRSSQCSRGLERYISQEEGPLSPFLGQLDEDYRTKETFLHRSDYSPH
100 μg
Chromatin Research
24149
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Western Blot, Immunohistochemistry
Unconjugated
PBS buffer, 2% sucrose
Rabbit
Affinity purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.