Learn More
ZNF318/TZF Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31 - $568.26
Specifications
Antigen | ZNF318/TZF |
---|---|
Applications | Immunofluorescence, Immunohistochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-157-1880
![]() |
Bethyl Laboratories, Inc
IHC00312T |
10 μL |
Each for $151.31
|
|
|||||
50-157-1879
![]() |
Bethyl Laboratories, Inc
IHC00312 |
100 μL |
Each for $568.26
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Orangutan, Gorilla and Chimpanzee
ZNF318 encodes a nuclear protein with a zinc finger motif of the Cys2-His2 type that is a novel corepressor of androgen receptor (AR).Specifications
ZNF318/TZF | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
endocrine regulator, endocrine regulatory protein, HRIHFB2436, testicular zinc finger, TZF, ZFP318 | |
ZNF318 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
Immunofluorescence, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Human | |
Q5VUA4 | |
24149 | |
Between 2049 and 2099 | |
Primary | |
4° C | |
ZNF318 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.