Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF319 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | ZNF319 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
ZNF319 Polyclonal specifically detects ZNF319 in Human samples. It is validated for Western Blot.Specifications
ZNF319 | |
Polyclonal | |
Rabbit | |
NP_065858 | |
57567 | |
Synthetic peptide directed towards the N terminal of human ZNF319The immunogen for this antibody is ZNF319. Peptide sequence TLPPGTAENPLGCAVYGILLQPDPGLQPPQHAPLQAAGEPGPKCGVCGHD. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
Zinc finger and SCAN domain-containing protein 15, zinc finger protein 397, Zinc finger protein 47ZSCAN15MGC13250, ZNF47 | |
ZNF319 | |
IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title