Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF320 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA584552
Description
Immunogen sequence: PIKGQLESRF HLHLRRHRRI.
ZNF320 encodes a Kruppel-like zinc finger protein. Members of this protein family are involved in activation or repression of transcription.
Specifications
ZNF320 | |
Polyclonal | |
Unconjugated | |
ZNF320 | |
DKFZp686G16228; ZFPL; zinc finger gene 320; zinc finger protein 320; zinc finger protein like; ZNF320 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
162967 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunofluorescence | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
A2RRD8 | |
ZNF320 | |
Recombinant protein corresponding to Human ZNF320. Recombinant protein control fragment (Product #RP-108473). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction