Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF322A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560262
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560262 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560262 Supplier Invitrogen™ Supplier No. PA560262
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QRIHMEEKPH QWSACESGFL LGMDFVAQQK MRTQTEELHY KYTVCD Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 91%.

ZNF322 contains four exons and spans 23.2kb in chromosome 6p22.1 region, and transcribes a 2.7kb mRNA that encodes a protein with 402 amino acid residues.Through northern blot analysis, ZNF322 was shown to be expressed in every human tissue examined at adult stage and during embryonic developmental stages from 80 days to 24 weeks. When ZNF322 was overexpressed in COS-7 cells, ZNF322-EGFP fusion protein is detected in the nucleus and cytoplasm. Reporter gene assays show that ZNF322 is a transcriptional activator. Furthermore, overexpression of ZNF322 in COS-7 cells activates the transcriptional activity of SRE and AP-1. Together, these results suggest that ZNF322 is a member of the zinc-finger transcription factor family and may act as a positive regulator in gene transcription mediated by the MAPK signaling pathways.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF322A
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF322
Gene Accession No. Q6U7Q0
Gene Alias HCG12; HLA complex group 12; Zfp322a; Zinc finger protein 322; zinc finger protein 322a; Zinc finger protein 388; zinc finger protein 489; ZNF322; ZNF322A; ZNF388; ZNF489
Gene Symbols ZNF322
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZNF322A. Recombinant protein control fragment (Product #RP-99502).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79692
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.