Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF323 Monoclonal Antibody (4F7)
GREENER_CHOICE

Catalog No. PIMA522484
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA522484 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA522484 Supplier Invitrogen™ Supplier No. MA522484
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: MEHLGDSKLQ RDVSLDSKYR ETCKRDSKAE KQQAHSTGER RHRCNECGKS FTKSSVLIEH QRIHTGEKPY ECEECGKAFS RRSSLNEHRR SHTGEKPYQC KECGKAFSAS NGLTRHRRIH TGEKPYECKV CGKAFLLSSC LVQHQRIHTG EKRYQCRECG KAFIQNTGLF QHLRVHTGEK PYQCSQCSKL FSKRTLLRKH QKIHTGERP.

ZNF323 is a member of the subfamily of C2H2 Kruppel-like zinc finger transcription factors that have a SCAN box domain (Pi et al., 2002 ).
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF323
Applications ELISA, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 4F7
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene ZSCAN31
Gene Accession No. Q96LW9
Gene Alias dJ874C20.2; zinc finger and SCAN domain containing 31; zinc finger and SCAN domain-containing protein 31; Zinc finger protein 323; ZNF20-Lp; ZNF310P; ZNF323; ZSCAN31
Gene Symbols ZSCAN31
Host Species Mouse
Immunogen ZSCAN31 (AAH08490, 1 a.a. ~ 209 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64288
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.