Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF323 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA552387
Description
Immunogen sequence: EEAVAVVEDL EQELSEPGNQ APDHEHGHSE VLLEDVEHLK VKQEPTDIQL QPMVTQLRYE SFCLHQFQEQ DGESIPENQE LASKQEILKE MEHLGDSKLQ RDVSLDSK Highest antigen sequence identity to the following orthologs: Mouse - 35%, Rat - 31%.
ZNF323 is a member of the subfamily of C2H2 Kruppel-like zinc finger transcription factors that have a SCAN box domain (Pi et al., 2002 ).
Specifications
ZNF323 | |
Polyclonal | |
Unconjugated | |
ZSCAN31 | |
dJ874C20.2; zinc finger and SCAN domain containing 31; zinc finger and SCAN domain-containing protein 31; Zinc finger protein 323; ZNF20-Lp; ZNF310P; ZNF323; ZSCAN31 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
64288 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96LW9 | |
ZSCAN31 | |
Recombinant protein corresponding to Human ZNF323. Recombinant protein control fragment (Product #RP-89443). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction